Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tekt2
Entrez Gene ID
298532
Gene Name
tektin 2 (testicular)
Gene Aliases
-
Chromosome Location
Chr.5: 144341955 - 144345494 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011977.1 NP_001011977.1 2 - 3 104 73
XM_006238860.1 XP_006238922.1 2 - 3 276 73
GenBank mRNA BC078990.1 - 2 - 3 104 73

View Full Product Details