Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tekt2
Entrez Gene ID
298532
Gene Name
tektin 2 (testicular)
Gene Aliases
-
Chromosome Location
Chr.5: 144341955 - 144345494 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011977.1 NP_001011977.1 7 - 8 905 60
XM_006238860.1 XP_006238922.1 7 - 8 1077 60
XM_006238861.2 XP_006238923.1 6 - 7 838 60
GenBank mRNA BC078990.1 - 7 - 8 905 60

View Full Product Details