The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106698.1 | NP_001100168.1 | 3 - 4 | 1031 | 62 |
XM_006239459.2 | XP_006239521.1 | 3 - 4 | 1181 | 62 | |
XM_006239460.2 | XP_006239522.1 | 4 - 5 | 1316 | 62 |