Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo44
Entrez Gene ID
500587
Gene Name
F-box protein 44
Gene Aliases
RGD1562463
Chromosome Location
Chr.5: 164962921 - 164972000 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006239395.2 XP_006239457.1 1 - 2 89 59

View Full Product Details