The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025018.1 | NP_001020189.1 | 9 - 10 | 1036 | 96 |
XM_006233120.2 | XP_006233182.1 | 9 - 10 | 980 | 96 | |
XM_006233121.2 | XP_006233183.1 | 8 - 9 | 920 | 96 | |
XM_006233122.2 | XP_006233184.1 | 8 - 9 | 1021 | 96 | |
XM_006233123.2 | XP_006233185.1 | 8 - 9 | 883 | 96 | |
GenBank mRNA | BC091251.1 | - | 9 - 10 | 1036 | 96 |
FQ218349.1 | - | 9 - 10 | 1048 | 96 | |
FQ228782.1 | - | 9 - 10 | 1133 | 96 |