Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ska1
Entrez Gene ID
291441
Gene Name
spindle and kinetochore associated complex subunit 1
Gene Aliases
RGD1310784
Chromosome Location
Chr.18: 70174316 - 70184368 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106134.1 NP_001099604.1 2 - 3 281 238
XM_006254918.2 XP_006254980.1 2 - 3 294 238
XM_006254920.2 XP_006254982.1 2 - 3 287 234
XM_008772106.1 XP_008770328.1 2 - 3 297 238
XM_008772107.1 XP_008770329.1 2 - 3 293 234
XM_008772108.1 XP_008770330.1 2 - 3 295 238
GenBank mRNA BC158659.1 - 2 - 3 233 238

View Full Product Details