The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108757.1 | NP_001102227.1 | 2 - 3 | 214 | 80 |
XM_008766061.1 | XP_008764283.1 | 1 - 2 | 348 | 80 |