Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arrdc5
Entrez Gene ID
680452
Gene Name
arrestin domain containing 5
Gene Aliases
-
Chromosome Location
Chr.9: 10760113 - 10766199 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109408.1 NP_001102878.1 1 - 2 248 142

View Full Product Details