Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Azgp1
Entrez Gene ID
25294
Gene Name
alpha-2-glycoprotein 1, zinc-binding
Gene Aliases
ZA2GA, Zna2gp
Chromosome Location
Chr.12: 19196563 - 19203094 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012826.2 NP_036958.2 1 - 2 79 60
XM_006249003.2 XP_006249065.1 1 - 2 122 60
XM_006249004.2 XP_006249066.1 1 - 2 121 60
GenBank mRNA BC105822.1 - 1 - 2 79 60
D21058.1 - 1 - 2 78 60
FQ210978.1 - - 98 60
FQ219509.1 - 1 - 2 70 60
X75309.1 - 1 - 2 60 60

View Full Product Details