The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012826.2 | NP_036958.2 | 2 - 3 | 347 | 126 |
GenBank mRNA | BC105822.1 | - | 2 - 3 | 347 | 126 |
D21058.1 | - | 2 - 3 | 346 | 126 | |
FQ210978.1 | - | - | 366 | 126 | |
FQ219509.1 | - | 2 - 3 | 338 | 126 | |
X75309.1 | - | 2 - 3 | 328 | 126 |