The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012566.1 | NP_036698.1 | 4 - 5 | 852 | 119 |
XM_008769917.1 | XP_008768139.1 | 4 - 5 | 1093 | 119 | |
XM_008769918.1 | XP_008768140.1 | 4 - 5 | 928 | 119 | |
XM_008769919.1 | XP_008768141.1 | 4 - 5 | 1810 | 119 | |
XM_008769920.1 | XP_008768142.1 | 4 - 5 | 1810 | 119 | |
GenBank mRNA | L06986.1 | - | 4 - 5 | 852 | 119 |