Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pou6f1
Entrez Gene ID
116545
Gene Name
POU class 6 homeobox 1
Gene Aliases
Brn5
Chromosome Location
Chr.7: 142201614 - 142225256 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105746.1 NP_001099216.1 2 - 3 1046 70
XM_006242298.2 XP_006242360.1 8 - 9 1709 70
XM_006242301.2 XP_006242363.1 7 - 8 1946 70
XM_008765654.1 XP_008763876.1 7 - 8 2060 70
XM_008765655.1 XP_008763877.1 8 - 9 1721 70
XM_008765656.1 XP_008763878.1 8 - 9 1779 70
XM_008765657.1 XP_008763879.1 7 - 8 2057 70
XM_008765658.1 XP_008763880.1 7 - 8 2048 70
GenBank mRNA L23204.1 - 3 - 4 1023 70

View Full Product Details