Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bpifa3
Entrez Gene ID
690402
Gene Name
BPI fold containing family A, member 3
Gene Aliases
Splunc3
Chromosome Location
Chr.3: 149606925 - 149616787 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109588.1 NP_001103058.1 6 - 7 877 64

View Full Product Details