Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpr97
Entrez Gene ID
291854
Gene Name
G protein-coupled receptor 97
Gene Aliases
-
Chromosome Location
Chr.19: 10392001 - 10422604 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006255126.2 XP_006255188.1 4 - 5 2515 151

View Full Product Details