Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gnao1
Entrez Gene ID
50664
Gene Name
guanine nucleotide binding protein (G protein), alpha activating activity polypeptide O
Gene Aliases
Gnao, RATBPGTPC
Chromosome Location
Chr.19: 11512114 - 11669586 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017327.1 NP_059023.1 2 - 3 861 117
XM_008772320.1 XP_008770542.1 2 - 3 870 117
XM_008772321.1 XP_008770543.1 2 - 3 870 117
GenBank mRNA M17526.1 - 2 - 3 861 117

View Full Product Details