Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aqp7
Entrez Gene ID
29171
Gene Name
aquaporin 7
Gene Aliases
-
Chromosome Location
Chr.5: 57358327 - 57372239 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019157.2 NP_062030.2 3 - 4 482 87
GenBank mRNA AB000507.2 - 3 - 4 482 87
AY120935.1 - 2 - 3 221 87
AY157737.1 - 2 - 3 221 87
BC072695.1 - 3 - 4 442 87

View Full Product Details