Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mef2d
Entrez Gene ID
81518
Gene Name
myocyte enhancer factor 2D
Gene Aliases
-
Chromosome Location
Chr.2: 187512164 - 187541285 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030860.2 NP_110487.2 9 - 10 1653 71
XM_006232771.1 XP_006232833.1 9 - 10 1252 71
XM_006232772.1 XP_006232834.1 9 - 10 1249 71
XM_006232773.1 XP_006232835.1 8 - 9 1228 71
GenBank mRNA AJ005425.1 - 8 - 9 1228 71
BC081790.1 - 9 - 10 1653 71

View Full Product Details