Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gzmc
Entrez Gene ID
171290
Gene Name
granzyme C
Gene Aliases
Rnpk-4, Rnpk4
Chromosome Location
Chr.15: 35392158 - 35394787 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134332.2 NP_599159.1 2 - 3 211 104
GenBank mRNA FQ219852.1 - 2 - 3 274 104
FQ219889.1 - 2 - 3 274 104
FQ219966.1 - 2 - 3 272 104
FQ219998.1 - 2 - 3 276 104
FQ220005.1 - 2 - 3 274 104
FQ220014.1 - 2 - 3 274 104
FQ220035.1 - 2 - 3 274 104
FQ220062.1 - 2 - 3 276 104
FQ220064.1 - 2 - 3 274 104
FQ220079.1 - 2 - 3 272 104
FQ220128.1 - 2 - 3 272 103
FQ220142.1 - 2 - 3 274 104
FQ220223.1 - 2 - 3 274 104
FQ220228.1 - 2 - 3 264 104
FQ220350.1 - 2 - 3 274 104
FQ220364.1 - 2 - 3 274 104
FQ220426.1 - 2 - 3 274 104
FQ220453.1 - 2 - 3 274 104
FQ220464.1 - 2 - 3 274 104
FQ220466.1 - 2 - 3 274 104
FQ220480.1 - 2 - 3 276 104
FQ220512.1 - 2 - 3 274 104
FQ220562.1 - 2 - 3 274 104
FQ220600.1 - 2 - 3 274 104
FQ220749.1 - 2 - 3 273 104
FQ220751.1 - 2 - 3 272 104
FQ220856.1 - 2 - 3 273 104
FQ220878.1 - 2 - 3 273 104
FQ220928.1 - 2 - 3 276 104
FQ221390.1 - 2 - 3 274 104
FQ221473.1 - 2 - 3 274 104
FQ221940.1 - 2 - 3 274 104
FQ223110.1 - 2 - 3 274 104
FQ228922.1 - 2 - 3 276 104
FQ229160.1 - 2 - 3 274 104
FQ229161.1 - 2 - 3 274 104
FQ229162.1 - 2 - 3 274 104
FQ229181.1 - 2 - 3 276 104
FQ229215.1 - 2 - 3 274 104
FQ229235.1 - 2 - 3 278 104
FQ229289.1 - 2 - 3 274 104
FQ229298.1 - 2 - 3 274 104
FQ229311.1 - 2 - 3 274 104
FQ229314.1 - 2 - 3 274 104
FQ229395.1 - 2 - 3 274 104
FQ229439.1 - 2 - 3 272 104
FQ229452.1 - 2 - 3 274 104
FQ229454.1 - 2 - 3 274 104
FQ229468.1 - 2 - 3 274 104
FQ229486.1 - 2 - 3 274 104
FQ229502.1 - 2 - 3 274 104
FQ229509.1 - 2 - 3 274 104
FQ229543.1 - 2 - 3 274 104
FQ229636.1 - 2 - 3 274 104
FQ229685.1 - 2 - 3 274 104
FQ229690.1 - 2 - 3 318 104
FQ229764.1 - 2 - 3 275 104
FQ229772.1 - 2 - 3 272 104
FQ229825.1 - 2 - 3 274 104
FQ229868.1 - 2 - 3 305 104
FQ229912.1 - 2 - 3 274 104
FQ229913.1 - 2 - 3 272 104
FQ229994.1 - 2 - 3 276 104
FQ230028.1 - 2 - 3 276 104
FQ230087.1 - 2 - 3 274 104
FQ230100.1 - 2 - 3 274 104
FQ230157.1 - 2 - 3 276 104
FQ230165.1 - 2 - 3 274 104
U57062.1 - 2 - 3 212 104

Gene Symbol
LOC691695
Entrez Gene ID
691695
Gene Name
similar to granzyme C
Gene Aliases
-
Chromosome Location
Chr.15: 36448970 - 36451720 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751488.3 XP_003751536.2 2 - 3 332 104
GenBank mRNA FQ219891.1 - 2 - 3 274 104
FQ229899.1 - 2 - 3 274 104

View Full Product Details