The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199371.1 | NP_955403.1 | 5 - 6 | 695 | 90 |
XM_006245922.2 | XP_006245984.1 | 6 - 7 | 886 | 90 | |
XM_006245923.2 | XP_006245985.1 | 6 - 7 | 894 | 90 | |
XM_008767553.1 | XP_008765775.1 | 6 - 7 | 914 | 90 | |
GenBank mRNA | AB073023.1 | - | 5 - 6 | 695 | 90 |