Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asb15
Entrez Gene ID
500050
Gene Name
ankyrin repeat and SOCS box containing 15
Gene Aliases
-
Chromosome Location
Chr.4: 51598829 - 51642276 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006236158.2 XP_006236220.1 4 - 5 644 71
XM_008762831.1 XP_008761053.1 4 - 5 800 71
XM_008762832.1 XP_008761054.1 4 - 5 601 71
XM_008762833.1 XP_008761055.1 4 - 5 822 71
GenBank mRNA BC097405.1 - 4 - 5 356 71

View Full Product Details