Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asb15
Entrez Gene ID
500050
Gene Name
ankyrin repeat and SOCS box containing 15
Gene Aliases
-
Chromosome Location
Chr.4: 51598829 - 51642276 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006236158.2 XP_006236220.1 5 - 6 797 116
XM_008762831.1 XP_008761053.1 5 - 6 953 116
XM_008762832.1 XP_008761054.1 5 - 6 754 116
XM_008762833.1 XP_008761055.1 5 - 6 975 116
GenBank mRNA BC097405.1 - 5 - 6 509 116

View Full Product Details