Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clec2l
Entrez Gene ID
296985
Gene Name
C-type lectin domain family 2, member L
Gene Aliases
RGD1565373
Chromosome Location
Chr.4: 66405166 - 66421654 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044233.2 NP_001037698.2 3 - 4 421 61
GenBank mRNA BC161812.1 - - 551 61
DQ630549.1 - 3 - 4 281 61

View Full Product Details