Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chia
Entrez Gene ID
113901
Gene Name
chitinase, acidic
Gene Aliases
-
Chromosome Location
Chr.2: 208750348 - 208797388 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA KC529649.1 - 5 - 6 414 81

View Full Product Details