Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chia
Entrez Gene ID
113901
Gene Name
chitinase, acidic
Gene Aliases
-
Chromosome Location
Chr.2: 208750348 - 208797388 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207586.2 NP_997469.1 2 - 3 73 78
GenBank mRNA AY486074.1 - 2 - 3 75 78

View Full Product Details