Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klrc1
Entrez Gene ID
29683
Gene Name
killer cell lectin-like receptor subfamily C, member 1
Gene Aliases
rNKG2A
Chromosome Location
Chr.4: 163453435 - 163463718 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF021350.1 - 3 - 4 330 134

View Full Product Details