Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snrnp40
Entrez Gene ID
313056
Gene Name
small nuclear ribonucleoprotein 40 (U5)
Gene Aliases
RGD1309198
Chromosome Location
Chr.5: 148577498 - 148610658 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134556.1 NP_001128028.1 7 - 8 900 134

View Full Product Details