The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107802.2 | NP_001101272.1 | 1 - 2 | 1212 | 61 |
GenBank mRNA | BC158617.1 | - | 1 - 2 | 1212 | 61 |