Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sarm1
Entrez Gene ID
287545
Gene Name
sterile alpha and TIR motif containing 1
Gene Aliases
-
Chromosome Location
Chr.10: 65742343 - 65766050 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105817.1 NP_001099287.1 4 - 5 1518 76

View Full Product Details