Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fads6
Entrez Gene ID
303671
Gene Name
fatty acid desaturase 6
Gene Aliases
RGD1311950
Chromosome Location
Chr.10: 103832838 - 103848035 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107064.1 NP_001100534.1 3 - 4 548 79

View Full Product Details