Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stac2
Entrez Gene ID
363674
Gene Name
SH3 and cysteine rich domain 2
Gene Aliases
-
Chromosome Location
Chr.10: 85988367 - 86004209 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108834.1 NP_001102304.1 1 - 2 570 80
XM_006247383.2 XP_006247445.1 1 - 2 461 80
GenBank mRNA AM903074.1 - 1 - 2 95 80

View Full Product Details