Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Itgbl1
Entrez Gene ID
498564
Gene Name
integrin, beta-like 1
Gene Aliases
-
Chromosome Location
Chr.15: 110114148 - 110378970 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017505.1 NP_001017505.1 3 - 4 558 70
GenBank mRNA BC087076.1 - 3 - 4 558 70

View Full Product Details