Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Actr3
Entrez Gene ID
81732
Gene Name
ARP3 actin-related protein 3 homolog (yeast)
Gene Aliases
Arp3
Chromosome Location
Chr.13: 41695520 - 41738622 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031068.1 NP_112330.1 6 - 7 627 121
GenBank mRNA AB292042.1 - 6 - 7 551 121
AB292043.1 - 6 - 7 540 121
BC098014.1 - 6 - 7 627 121
FQ212256.1 - 6 - 7 829 121

View Full Product Details