Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hdx
Entrez Gene ID
317617
Gene Name
highly divergent homeobox
Gene Aliases
RGD1563666
Chromosome Location
Chr.X: 82847895 - 83151516 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134568.1 NP_001128040.1 - 245 108

View Full Product Details