Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dctn5
Entrez Gene ID
308961
Gene Name
dynactin 5 (p25)
Gene Aliases
RGD1305654
Chromosome Location
Chr.1: 192088709 - 192105421 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037778.1 NP_001032867.1 2 - 3 159 84
GenBank mRNA BC098764.1 - 2 - 3 159 84

View Full Product Details