The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012238.1 | NP_001012238.1 | 9 - 10 | 2008 | 65 |
GenBank mRNA | BC161825.1 | - | 9 - 10 | 1675 | 65 |