Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhobtb3
Entrez Gene ID
309922
Gene Name
Rho-related BTB domain containing 3
Gene Aliases
-
Chromosome Location
Chr.2: 2627655 - 2683305 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107645.1 NP_001101115.1 6 - 7 1467 84

View Full Product Details