The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135875.1 | NP_001129347.1 | 10 - 11 | 1522 | 60 |
XM_008772517.1 | XP_008770739.1 | 9 - 10 | 1537 | 60 |