The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100836.1 | NP_001094306.1 | 5 - 6 | 857 | 139 |
GenBank mRNA | BC158809.1 | - | 5 - 6 | 858 | 139 |