Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfp74
Entrez Gene ID
365224
Gene Name
zinc finger protein 74
Gene Aliases
Znf569
Chromosome Location
Chr.1: 88231016 - 88257564 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271339.1 NP_001258268.1 3 - 4 394 86
XM_006228731.2 XP_006228793.1 4 - 5 1543 86
XM_006228732.2 XP_006228794.1 4 - 5 1538 86

Gene Symbol
LOC103690038
Entrez Gene ID
103690038
Gene Name
zinc finger protein 569
Gene Aliases
-
Chromosome Location
Chr.1: 87816292 - 87842539 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006228711.2 XP_006228773.1 4 - 5 1543 86
XM_006228712.2 XP_006228774.1 4 - 5 1538 86

View Full Product Details