Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp74
Entrez Gene ID
365224
Gene Name
zinc finger protein 74
Gene Aliases
Znf569
Chromosome Location
Chr.1: 88231016 - 88257564 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271339.1 NP_001258268.1 4 - 5 485 96
XM_006228731.2 XP_006228793.1 5 - 6 1634 96
XM_006228732.2 XP_006228794.1 5 - 6 1629 96

Gene Symbol
LOC103690038
Entrez Gene ID
103690038
Gene Name
zinc finger protein 569
Gene Aliases
-
Chromosome Location
Chr.1: 87816292 - 87842539 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006228711.2 XP_006228773.1 5 - 6 1634 96
XM_006228712.2 XP_006228774.1 5 - 6 1629 96

View Full Product Details