Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptdss1
Entrez Gene ID
314553
Gene Name
phosphatidylserine synthase 1
Gene Aliases
PSS-1
Chromosome Location
Chr.7: 71294140 - 71356153 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012113.1 NP_001012113.1 8 - 9 1111 69
GenBank mRNA BC087129.1 - 8 - 9 1111 69

View Full Product Details