Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhdc1
Entrez Gene ID
314190
Gene Name
kelch domain containing 1
Gene Aliases
-
Chromosome Location
Chr.6: 91532380 - 91590998 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108027.1 NP_001101497.1 2 - 3 278 72
XM_006240169.2 XP_006240231.1 2 - 3 183 72
XM_006240170.2 XP_006240232.1 2 - 3 177 72
XM_008764716.1 XP_008762938.1 2 - 3 183 72
XM_008764717.1 XP_008762939.1 2 - 3 91 72
GenBank mRNA BC159403.1 - 2 - 3 189 72

View Full Product Details