Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sun3
Entrez Gene ID
498412
Gene Name
Sad1 and UNC84 domain containing 3
Gene Aliases
Sunc1
Chromosome Location
Chr.14: 89215412 - 89253581 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006251471.2 XP_006251533.1 5 - 6 666 83
XM_008770406.1 XP_008768628.1 5 - 6 4248 83

View Full Product Details