Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Xlr4a
Entrez Gene ID
293841
Gene Name
X-linked lymphocyte-regulated 4A
Gene Aliases
RGD1565442
Chromosome Location
Chr.X: 153072965 - 153081322 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006229523.2 XP_006229585.1 6 - 7 993 150
XM_006229524.2 XP_006229586.1 6 - 7 1008 150
XM_006229527.2 XP_006229589.1 6 - 7 613 150
XM_006229528.2 XP_006229590.1 6 - 7 975 150
XM_006229529.2 XP_006229591.1 6 - 7 990 150
XM_219738.8 XP_219738.6 5 - 6 1152 150

View Full Product Details