Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vom2r38
Entrez Gene ID
308551
Gene Name
vomeronasal 2 receptor, 38
Gene Aliases
RGD1565669
Chromosome Location
Chr.1: 98915864 - 99145996 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099476.1 NP_001092946.1 - 505 116

View Full Product Details