Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Frg1
Entrez Gene ID
686103
Gene Name
FSHD region gene 1
Gene Aliases
-
Chromosome Location
Chr.16: 53957858 - 53978865 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290133.1 NP_001277062.1 8 - 9 874 140

View Full Product Details