Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubr7
Entrez Gene ID
314399
Gene Name
ubiquitin protein ligase E3 component n-recognin 7 (putative)
Gene Aliases
RGD1359144
Chromosome Location
Chr.6: 126636606 - 126656513 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007705.1 NP_001007706.1 11 - 11 2797 89
XM_006240475.2 XP_006240537.1 12 - 12 2913 89
XM_006240476.2 XP_006240538.1 12 - 12 2910 89
GenBank mRNA BC081966.1 - 11 - 11 2797 89

View Full Product Details