Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Mtif2
Entrez Gene ID
305606
Gene Name
mitochondrial translational initiation factor 2
Gene Aliases
-
Chromosome Location
Chr.14: 113946084 - 113965574 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004254.1 NP_001004254.1 11 - 11 1597 147
GenBank mRNA BC079421.1 - 11 - 11 1597 147

View Full Product Details