The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134750.1 | NP_001128222.1 | 10 - 10 | 2821 | 129 |
NM_001134751.1 | NP_001128223.1 | 9 - 9 | 2739 | 129 | |
GenBank mRNA | BC168228.1 | - | 9 - 9 | 3004 | 129 |