Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ly49i7
Entrez Gene ID
494210
Gene Name
immunoreceptor Ly49i7
Gene Aliases
-
Chromosome Location
Chr.4: 162712736 - 162733947 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009500.1 NP_001009500.1 2 - 3 198 138
XM_008763320.1 XP_008761542.1 3 - 4 1295 138
GenBank mRNA AY659934.1 - 2 - 3 198 138

Gene Symbol
LOC100911272
Entrez Gene ID
100911272
Gene Name
killer cell lectin-like receptor 5-like
Gene Aliases
-
Chromosome Location
Chr.4: 165347731 - 165366543 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008763440.1 XP_008761662.1 3 - 4 1054 138

View Full Product Details